Drukuj
Odczynnik do transdukcji, ViroMag™ R/L
Dostawca: OZ BIOSCIENCES
RL40100 RL40200 RL41000 KC30700
OZBIRL40100EA 592 PLN
OZBIRL40100 OZBIRL40200 OZBIRL41000 OZBIKC30700
Odczynnik do transdukcji, ViroMag™ R/L
Odczynniki do transfekcji
ViroMag™ R/L transduction reagent is a magnetic nanoparticles formulation optimised for rRetroviruses and lentiviruses. Based on Magnetofection™ technology, this reagent allows concentrating the complete applied dose of retro/lentiviral particles onto cells within minutes, inducing a significant improvement of virus infectivity with extremely low vector doses.
  • Increase transduction efficiency
  • Accelerate the transduction process
  • Efficient for hard-to-infect and non-permissive cells
  • Concentrate the viral dose onto the cells
  • Suitable for all type of retro/lentivirus
  • Synchronise cells adsorption/infection without modifications of the viruses
ViroMag™ R/L is ideal to infect non permissive cells. This reagent demonstrates an exceptionally high efficiency to promote, control and assist viral transductions in a so that no molecular biology processes or biochemical modifications are required.
Store at +4 °C.
Informacje do zamówienia: Needs to be used with a magnetic plate, available as a separate accessory or as part of a starter kit.
Dodatkowe informacje: Shipped at room temperature.