Drukuj
Zestawy magnetyczne
Dostawca: OZ BIOSCIENCES
IV-MAG1 IV-MAG2 IV-MAG3
OZBIIV-MAG1EA 344 PLN
OZBIIV-MAG1 OZBIIV-MAG2 OZBIIV-MAG3
Zestawy magnetyczne
Magnesy
These magnetic sets have been developped for efficient in vivo transfection or transduction with in vivo Magnetofection technology.
  • Can be easily cleaned and decontaminated with 70% ethanol, or by dry or wet sterilisation
  • Can be used at room temperature, 37 °C, +4 °C, etc.
  • Magnetic properties, distance between magnets and cells and incubation time have been optimized to efficiently concentrate nucleic acids or virus at the targetd site
  • Solid, completely reusable, it is a one-time buy
Magnetic properties, distance between magnets and cells andWięcej informacji o produkcie