Drukuj
Odczynnik do transfekcji DNA, HeLaFect™
Dostawca: OZ BIOSCIENCES
HF20500 HF21000
OZBIHF20500EA 847 PLN
OZBIHF20500 OZBIHF21000
Odczynnik do transfekcji DNA, HeLaFect™
Odczynniki do transfekcji
HeLaFect™ transfection reagent is specifically developed for HeLa cell lineage transfection with high efficiency. It is a lipid-based reagent based on Tee-Technology (Triggered Endosomal Escape). The cationic design of the reagent allows high nucleic acid compaction for efficient transport into HeLa cells. This reagent is composed of lipids, leading to high viability, and is ready to use.
  • Highly efficient, more than 80% of transfected HeLa cells
  • Ready-to-use, no need for additional buffer
  • Low nucleic acid amount minimised toxicity
  • High level of nucleic acid compaction
  • Easy and straight forward protocol
  • Compatible with any culture medium, medium change not required
Store at –20°C.
Dodatkowe informacje: Shipped at room temperature.
Zobacz ceny
SPECYFIKACJE
Specyficzne dla komórki HeLa
Environmentally Preferable