Drukuj
Kompletny adiuwant Freunda, CFAVax
Dostawca: OZ BIOSCIENCES
CFA0010
OZBICFA0010EA 308 PLN
OZBICFA0010
Kompletny adiuwant Freunda, CFAVax
Ekspresji białek Odczynniki do ekspresji białek
Complete Freund’s Adjuvant (CFA) is a water-in-oil emulsion containing 1 mg per ml heat-killed Mycobacterium tuberculosis.
  • Suitable for the stimulation of Th1 response, initial immunisation and antibody production
It consists of a mixture of mineral oil and emulsifier in a ratio of 85% v/v oil and 15% v/v emulsifier. It is not a pre-formed emulsion and thus it must be mixed with an equal volume of aqueous solution of antigen and subsequently emulsified prior to use. The emulsifier is mannide monooleate, an ester consisting of mannitol as the hydrophilic residue and oleic acid (unsaturated C18 fatty acid) as the hydrophobic part.
Store at +4 °C
Dodatkowe informacje: Shipped at room temperature.
Uwaga: Intended for research use only.