Drukuj
Human Recombinant OGG1 (from E. coli)
Dostawca: Novus Biologicals
NBP1-45318-0.1
NOVUNBP1-45318-0.1EA 0 PLN
NOVUNBP1-45318-0.1
Human Recombinant OGG1 (from E. coli)
Białka
OGG1, also known as 8-oxoguanine glycosylase, is a DNA glycosylase enzyme involved in base excision repair. This protein is the primary enzyme responsible for the excision of 7,8-dihydro-8-oxoguanine (8-oxoG), a mutagenic base byproduct which occurs as a result of exposure to reactive oxygen species (ROS).
It has a beta lyase activity that nicks DNA 3' to the lesion. Recombinant human OGG1 protein, fused to His-tag at N-terminus, was expressed in E.coli and purified by using conventional chromatography.
Uwaga: This product is for research use only and is not approved for use in humans or in clinical diagnosis.