Drukuj
PIG® SpillBlocker® Dike, chropowata powierzchnia
Dostawca: NEW PIG
PLR230
NEWPPLR230EA 2040 PLN
NEWPPLR230
PIG® SpillBlocker® Dike, chropowata powierzchnia
Materiały do kontroli wycieków
Spills on asphalt can seep underneath standard spill dikes. Stop them in their tracks with our rough surface dike.
  • Exclusive New Pig formulation features a tear-resistant top layer with a super-tacky bottom layer that seals tight to rough surfaces and blocks liquids
  • Bottom layer "seeps" into surface cracks to form a liquid-tight seal
  • Fast protection for when a spill is heading right for your drain on a rough surface; just set dike down with sticky side away from surface, turn dike over and press into place
  • Confines and diverts fluids without absorbing them,