Drukuj
Zwijana paleta do rozlewania w twardej oprawie, PIG®
Dostawca: NEW PIG
PAK601-AWO PAK601-BWG-AWO
NEWPPAK601-AWOEA 8620 PLN
NEWPPAK601-AWO NEWPPAK601-BWG-AWO
Zwijana paleta do rozlewania w twardej oprawie, PIG®
Składowanie beczek
These roll top hardcovers can be used for all-weather protection and liquid containment. Durable swing-out doors on front offers easy loading and unloading.
  • Weather-resistant design prevents contaminants from entering stormwater
  • Heavy-duty construction allows hardcover and pallet to be lifted by a forklift with drums in place
  • Low-density polyethylene (LDPE) with UV inhibitors resists UV rays, rust, corrosion and most chemicals for long, trouble-free life
  • Doors and roll top are available with a padlock