Drukuj
Maty i rolki absorbujące PIG Blue®
Dostawca: NEW PIG
BLU101 BLU103 BLU100
NEWPBLU101EA 782 PLN
NEWPBLU101 NEWPBLU103 NEWPBLU100
Maty i rolki absorbujące PIG Blue®
Materiały do kontroli wycieków
PIG Blue is the most absorbent mat. Maximum absorbency soaks up more so you use less on big spills and high-volume leaks.
  • Consistent construction produces a mat that's free of light spots to absorb liquid evenly for better wicking and retention
  • FibreFusion Technology fuses superabsorbent natural fibres with strong synthetic fibres for one high-performance absorbent
  • Stays together, even when picking up or shifting a fully saturated mat
  • Contains 90% recycled fibres; ideal in situations where green products are desired or required
  • Fire retardant for added
Więcej informacji o produkcie