Drukuj
Centrifuge Tube with Attachment
Dostawca: NEUBERT VOLUME GLASSWAERE
0459-00-0054
NEUB0459-00-0054EA 371 PLN
NEUB0459-00-0054
Centrifuge Tube with Attachment
Probówki Probówki wirówkowe
The centrifuge tube is made up of borosilicate 3.3 glass.
  • Content 10 ml ring mark: 100% content
  • Straight edge
  • Approch: 0 to 0,4 ml in 0,01 ml