Drukuj
Czarne worki, Whirl-Pak®
Dostawca: NASCO FORT ATKINSON
B01418WA B01472WA B01558WA
NASCB01418WAEA 2360 PLN
NASCB01418WA NASCB01472WA NASCB01558WA
Czarne worki, Whirl-Pak®
Worki Woreczki na próbki
Your solution for light sensitive samples - an opaque bag that provides maximum protection for samples that are sensitive to light.
  • Plus 1 sterilisation, post-manufacturing sterilisation: Increased sample integrity, perforated closure to maintain sterility
  • PREMIUM opaque film, keep your light sensitive samples safe
  • Convenient write-on strip for easy labeling
  • Puncture-proof closure tabs: Inner and outer tapes for longevity and stability, wire for easy opening, pouring, and closing
  • Opening pull tabs: For easy opening, non-slip textured Tyvek
  • Continuous
Więcej informacji o produkcie
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable