Drukuj
Kołnierz nagłowia do respiratorów powietrznych
Dostawca: 3M
M444
MMMAM444EA 437 PLN
MMMAM444
Kołnierz nagłowia do respiratorów powietrznych
Ochrona głowy Kaski ochronne
Replacement inner collar for use with the Versaflo™ M-400 Headtop. The inner collar zips into the shroud and provides a comfortable respiratory seal.
  • Replacement inner collar
  • Provides a comfortable respiratory seal
  • Made from polypropylene coated knitted polyamid
Replacement inner collar is designed for use with the 3M™Versaflo™ M-400 Series Helmets.