Drukuj
Dozowniki cieczy i inne akcesoria
Dostawca: MIELE
69.7470.06 69.7470.08 3838110 7514610 7514600 6425780 6425750 4518050 10087330 10087340
MIEL69.7470.06EA 0 PLN
MIEL69.7470.06 141-0138 141-0391 141-0579 141-0578 141-0513 141-0512 141-0395 MIEL10087330 MIEL10087340
Dozowniki cieczy i inne akcesoria
Zmywarki laboratoryjne
G 7896 DOS cabinet: for 1 to 4 DOS modules with containers.
  • Removable door
  • Freestanding, can be built under a work plan
  • Casing stainless steel or white steel
  • Three levels: one level with pull-out drawer on telescopic runnders for the storage of up 4 DOS modules, the levels 2 and 3 with pull-out drawer on telescopic runners sith drip tray and retainer for storage of 2×5 litre containers
DOS G 60 module:
For liquid cleaning agents
Siphon dispensing pump, electronically controlled by the machine
333 mm siphon with magnetic float to indicate when 5 and 10Więcej informacji o produkcie
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable