Drukuj
Rurki przesyłowe Masterflex®, Norprene® Pressure, Avantor®
Dostawca: Avantor Fluid Handling
95715-00
MFLX95715-00EA 2110 PLN
MFLX95715-00
Rurki przesyłowe Masterflex®, Norprene® Pressure, Avantor®
Przewody, kapilary i rury Przewody rurowe
Flexibility at extreme temperatures.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Ideal for chemical transfer, pesticide/fertilizer delivery, water feed lines, food/dairy processing, and water purification lines. The tube features excellent resistance to acidic and alkali substances, ozone, UV light, andWięcej informacji o produkcie