Drukuj
Analogowe napędy konsolowe o zmiennej prędkości Masterflex® L/S®, Avantor®
Dostawca: Avantor Fluid Handling
MFLX77910-45-EUEA 11090 PLN
MFLX77910-45-EU MFLX77910-55-EU MFLX77910-65-EU MFLX77910-75-EU
Analogowe napędy konsolowe o zmiennej prędkości Masterflex® L/S®, Avantor®
Pompy Pompy perystaltyczne
Compact, cost effective pump systems for general fluid transfer applications.
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Reversible motor lets you easily prime/purge tubing and pump in either direction
  • Remote control of start/stop via DB9 connector on back of drive
Choose a convenient, complete system all with one catalog number.