Drukuj
Systemy pomp o zmiennej prędkości obrotowej Masterflex® Ismatec® Microflex, Avantor®
Dostawca: Avantor Fluid Handling
MFLX77122-30-EUEA 6150 PLN
MFLX77122-30-EU MFLX77122-32-EU MFLX77122-34-EU
Systemy pomp o zmiennej prędkości obrotowej Masterflex® Ismatec® Microflex, Avantor®
Pompy Pompy perystaltyczne
Easy-to-use low-flow pumps feature reversible pumping, remote start/stop, and rapid tubing changes.
  • Polyphenylene sulfide (PPS), acetal, and copolyester pump head with four stainless steel (SS) rollers
  • Pumps accept continuous lengths of tubing for a clean, uninterrupted flow path with no fittings.
  • Separate single-turn speed control and cw/off/ccw switch with green LED power indicator—maintain speed setting when you turn pump on/off or reverse direction.
  • Reversible permanent magnet DC motor; purge fluid before or after pumping; pump fluid in either
Więcej informacji o produkcie