Drukuj
Przetwornice analogowe o zmiennej prędkości i zasilane powietrzem Masterflex® B/T®, Avantor®
Dostawca: Avantor Fluid Handling
77111-60
MFLX77111-60EA 34400 PLN
MFLX77111-60 MFLX77111-80 MFLX77711-95 MFLX77111-67-EU
Przetwornice analogowe o zmiennej prędkości i zasilane powietrzem Masterflex® B/T®, Avantor®
Pompy Pompy perystaltyczne
High-volume transfer and dispensing.
  • Flow rate 0.71/2.1 to 42 LPM with B/T tubing—flow rate depends on drive rpm and tubing size
  • Rapid-Load® pump head occlusion mechanism slides occlusion bed out of the way for easy tube loading and improved tubing occlusion
  • Accepts B/T PerfectPosition™ pump tubing sizes B/T 87 and B/T 91
  • Interlock switch shuts off power to stop drive when door is opened
Masterflex® B/T® drives are the best choice for high-flow bulk-transfer applications. Their durable construction ensures long service life with minimal maintenance.Więcej informacji o produkcie