Drukuj
Pompy odśrodkowe z wirnikiem otwartym Masterflex®, Avantor®
Dostawca: Avantor Fluid Handling
75506-07 75506-09 75506-05
MFLX75506-07EA 7540 PLN
MFLX75506-07 MFLX75506-09 MFLX75506-05
Pompy odśrodkowe z wirnikiem otwartym Masterflex®, Avantor®
Pompy Pompy odśrodkowe
Suitable for suspended solids.
  • Close-coupled design offers smallest possible footprint to save space
  • Back-pullout construction allows for servicing of pump without disturbing inlet and outlet piping
  • Top centerline discharge ensures self-venting of entrapped gases
Typical applications include food process, cooling systems, beverage processing and pharmaceutical services. The open impeller design means the pump can easily handle suspended solids up to a 3/4" spherical shape. Pumps are non-overloading and rated for continuous duty throughout entireWięcej informacji o produkcie