Drukuj
Przepływomierze masowe powietrza/N2 z metalowymi korpusami
Dostawca: Avantor Fluid Handling
32648-10 32648-00 32648-04 32648-08 32648-14 32648-16 32648-19 32648-34 32648-36 32648-42 32648-45 32648-66 32659-00
MFLX32648-10EA 3480 PLN
MFLX32648-10 MFLX32648-00 MFLX32648-04 MFLX32648-08 MFLX32648-14 MFLX32648-16 MFLX32648-19 MFLX32648-34 MFLX32648-36 MFLX32648-42 MFLX32648-45 MFLX32648-66 MFLX32659-00
Przepływomierze masowe powietrza/N2 z metalowymi korpusami
Przepływomierze
Precisely measure flow rates and totals.
  • Proven design uses metal body components for added durability
  • Accumulated flow can be logged using an optional totalizer
  • Includes NIST-traceable certificate
These flowmeters feature an advanced U-shaped sensor that ensures accurate and repeatable results. Gas flow measurements are unaffected by moderate temperature and pressure variations at the inlet. The meter also allows a four-point calibration across the flow range to improve meter linearity. Output data from the meter can be sent via a 0 to 5 VDC or 4 to 2Więcej informacji o produkcie