Drukuj
Siatki druciane do sitek sanitarnych
Dostawca: Avantor Fluid Handling
29590-31 29590-33 29590-35 29590-36 29590-37 29590-39
MFLX29590-31EA 792 PLN
MFLX29590-31 MFLX29590-33 MFLX29590-35 MFLX29590-36 MFLX29590-37 MFLX29590-39
Siatki druciane do sitek sanitarnych
Filtry Sitka
Protect products and expensive process equipment from coarse or fine particulates.
  • Mesh overscreens are available in a variety of mesh sizes to meet your specific application
  • Overscreens are hemmed smooth with no stray wire ends
  • Overscreens fit both in-line and side-inlet sanitary strainer assemblies
Sanitary strainers are ideal for particulate removal or pre-filtration in the processing of foods, beverages, pharmaceuticals, cosmetics, and other critical compounds. Housings are available in in-line and side-inlet configurations.