Drukuj
Wielofunkcyjne odśrodkowe separatory/oddzielacze jazowe
Dostawca: Avantor Fluid Handling
17315-20 17315-21 17315-22 17315-23 17315-24
MFLX17315-20EA 193 PLN
MFLX17315-20 MFLX17315-21 MFLX17315-22 MFLX17315-23 MFLX17315-24
Wielofunkcyjne odśrodkowe separatory/oddzielacze jazowe
Pompy Separatory odśrodkowe
Mix, react, and/or separate liquid-liquid mixtures in a single device.
  • Highly flexible design operates effectively under changing inlet concentrations, variable flow rates, and flow disruptions
  • Interchangeable weirs allow for broad applications in research, pilot plant, or production operations
Weirs are available in various diameters. Use the large diameter weirs in applications with high-viscosity liquids.