Drukuj
Standardowe napędy cyfrowe Masterflex® L/S®, Avantor®
Dostawca: Avantor Fluid Handling
MFLX07522-20-EUEA 17620 PLN
MFLX07522-20-EU MFLX07522-30-EU
Standardowe napędy cyfrowe Masterflex® L/S®, Avantor®
Pompy Napędy pomp Napędy pomp perystaltycznych
Designed with maintenance-free high-accuracy motor and a graphical interface—makes setup and operation easier than ever!
  • Flow rate 0.001/0.006 to 566/3400 ml/min with L/S tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating modes—continuous run, timed dispense, copy dispense, or volume dispense
  • Select your preferred language from the user-friendly menu for programming and operation
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles;