Drukuj
Rurki do pomp precyzyjnych Masterflex® I/P®, C-Flex®, Avantor®
Dostawca: Avantor Fluid Handling
MFLX06424-26EA 1020 PLN
MFLX06424-26 MFLX06424-73 MFLX06424-82
Rurki do pomp precyzyjnych Masterflex® I/P®, C-Flex®, Avantor®
Przewody, kapilary i rury Przewody pompy perystaltycznej
Maintain flexibility at extreme temperatures.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory Approvals to match your unique application
  • Ultra smooth inner surface results in fewer extractables