Drukuj
Systemy regałowe, wózek na kółkach z solidnymi półkami, Super Erecta®
Dostawca: Metro International
F556EG
METOF556EGEA 7100 PLN
METOF556EG
Systemy regałowe, wózek na kółkach z solidnymi półkami, Super Erecta®
Meble Regały i szafki
The system consists of four solid shelves, posts, plastic split sleeves, donut bumpers and 127 mm poly castors (2 swivel; 2 brake). The raised 'ship's edge' around perimeter helps to contain spillage.
  • Chrome-plated posts with four galvanised flat shelves