Drukuj
Paski testowe, ozon
Dostawca: MACHEREY-NAGEL
90736
MANA90736EA 106 PLN
MANA90736
Paski testowe, ozon
Zestawy do testów środowiskowych
These Ozone test strips are designed for quick, convenient and reliable on-site Ozone testing. The easy procedure yields accurate testing results within ten minutes.
  • Results within minutes: Just expose to air, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
Ozone is a colourless toxic gas. It is irritating to eyes and mucous membranes and can cause respiratory complaints. With Ozone concentrations increasing world-wide, especially in the summer time, Ozone testing becomes more and more important.
An Ozone content of 180 μg/m³ in the air is considered as critical value above which sensitive persons should avoid strong physical exertion. The Ozone values published in the media only represent the Ozone concentrations in the vicinity of the measuring stations. However, actual Ozone values in other locations can deviate considerably from these published values due to meteorological and chemical influences. The reasons for such deviations are for wind, solar radiation, exhaust gases from vehicles and so on. For this reason an on-site measurement of Ozone concentrations is recommended.
Colour reaction: White to brown