Drukuj
Fiolki z zakręcaną szyjką, N 15
Dostawca: MACHEREY-NAGEL
702311 702096 70285 702097
MANA702311EA 156 PLN
MANA702311 MANA702096 MANA70285 MANA702097
Fiolki z zakręcaną szyjką, N 15
Vialki
These vials are used for storage of liquid samples.
  • Made of clear and amber glass
  • Flat bottom
  • 15-425 thread
  • Usable volume of 8 and 12 ml
The suitable screw closures have bonded septa.
- Septa are firmly connected with the cap (bonded)
- High quality silicone/PTFE septum materials