Drukuj
Fiolki EPA z zakręcaną szyjką, N 24
Dostawca: MACHEREY-NAGEL
702022 702021 702024 702023 702133 702132 702131 702074 702831 702865 702877 702884
MANA702022EA 240 PLN
MANA702022 MANA702021 MANA702024 MANA702023 MANA702133 MANA702132 MANA702131 MANA702074 MANA702831 MANA702865 MANA702877 MANA702884
Fiolki EPA z zakręcaną szyjką, N 24
Vialki
These screw neck vials are preferential used for VOC and TOC analyses.
  • Made of clear and amber glass
  • Available with flat and round bottom
Can be used on autosamplers for VOC and TOC analyses of Agilent (AQUATek 70, SOLATek 72, Archon Purge+Trap), Dionex (Thermo Scientific - ASE 200, ASE 350, AS-HV), PerkinElmer (AQUATek 100), Shimadzu (ASI-L, ASI-V), Thermo Scientific (HiPerTOC), and Varian (Agilent - Archon).
With closed top of screw closures the vials are used for sample storage.
Bonded and ready assembled screw closures are available forWięcej informacji o produkcie