Drukuj
Pepstatyna A, klasa proteomiki
Dostawca: VWR Chemicals
M183-5MGEA 1580 PLN
M183-5MG
Pepstatyna A, klasa proteomiki
Białka
An aspartic protease inhibitor of pepsin, renin, and cathepsin D. Can also block HIV protease. Not soluble in water.
FTIR: Suitable
Melting point: 220-240 ° C
Optical rotation (0.5%, MeOH): -95 to -85 °
Purity (HPLC): 75%