Drukuj
WHEATON® Lyophilisation Stopper, Bromobutyl
Dostawca: DWK Life Sciences
700004
LSUK700004EA 880 PLN
LSUK700004
WHEATON® Lyophilisation Stopper, Bromobutyl
Korki Korki gumowe
Igloo lyophilisation style stoppers.
  • Manufactured from the highest quality bromobutyl rubber formulation
  • Components manufactured to yield exceptional dimensional stability
Food safe (EU only).
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable