Drukuj
Zestawy testowe, zasadowość (dyski), CHECKIT®
Dostawca: Lovibond
146450
LOVI146450EA 156 PLN
LOVI146450
Zestawy testowe, zasadowość (dyski), CHECKIT®
Zestawy do testów środowiskowych
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work. Supplied with a generous number of different colour scales, it provides the basis for a comprehensive, easy-to use colorimetric analysis system.
  • Enables the use of large path lengths
  • The mirror optics makes use of the view through the entire length of the cell
  • Easy operation and exact reagent dosing
  • Tablet reagents have minimum guaranteed shelf life of 5/10 years
CHECKIT® disc contains a continuous colour scale which makes it possible to achieve an exact colour match between the colour standard and the sample. These discs are specially manufactured in selected materials to remain colour-stability over a long period and guarantee reliable, reproducible measurement results.
Dodatkowe informacje: Supplied with instruction manuals explaining the various stages of analysis.