Drukuj
Filmy do żeli agarozowych, GelBond®
Dostawca: LONZA
53761 53734 53740 53745 53746 53748 53749 53750 53759 53780
LONZ53761EA 8960 PLN
LONZ53761 LONZ53734 LONZ53740 LONZ53745 LONZ53746 LONZ53748 LONZ53749 LONZ53750 LONZ53759 LONZ53780
Filmy do żeli agarozowych, GelBond®
Plomby Filmy i folie uszczelniające
GelBond® film is a transparent, flexible polyester film designed to support agarose gels. Gels cast on GelBond® film remain permanently attached to the film through electrophoresis or immunodiffusion and all subsequent fixing, staining, destaining, and drying procedures. GelBond® Film is available either as precut sheets or rolls.
  • Agarose gels retain their original dimensions during staining and after drying
  • Gels, particularly thin ones, are easier to handle during staining, destaining, and drying when supported
  • Gel orientation can be recorded directly on
Więcej informacji o produkcie