Drukuj
Komora Laminarna PCR, Purifier®
Dostawca: LABCONCO
3970325
LBCP3970325EA 0 PLN
LBCP3970325
Komora Laminarna PCR, Purifier®
Dygestoria i komory Komory do PCR
Filtered PCR Enclosures direct HEPA-filtered air downward over the work area to provide ISO Class 5 conditions and protect your work from particulate contamination. These specialized vertical clean benches provide a controlled environment in which to perform polymerase chain reaction experiments. These enclosures include a variable digital timer and when the set time expires, the UV light automatically switches off in preparation for the next experiment.
  • Nominal downflow velocity of 45 to 65 fpm with aerodynamic air foil and disposible prefilter
  • UV-