Drukuj
Chemoodporne osłony na buty i obuwie, KLEENGUARD* A40
Dostawca: KIMBERLY CLARK
98710 98820 98700 98720 98800
KIMB98710EA 2820 PLN
KIMB98710 KIMB98820 KIMB98700 KIMB98720 KIMB98800
Chemoodporne osłony na buty i obuwie, KLEENGUARD* A40
Odzież jednorazowa
These disposable shoe and boot covers are made from anti-static film laminate with liquid splash and dust barrier properties. The covers improve safety and comfort in high-risk areas and help to reduce contamination in food processing and light industrial areas.
  • Protect against water-based splashes, liquids or dust particles
  • With or without slip-resistant PVC sole
  • Breathable, lightweight and non absorbent
  • Elasticated tops/ankles for secure fitting
One size fits all.