Drukuj

Chusteczki toaletowe, maxi jumbo, Scott®

Dostawca: KIMBERLY CLARK
8511
KIMB8511EA 615 PLN
KIMB8511
Chusteczki toaletowe, maxi jumbo, Scott®
Chusteczki Papier toaletowy
For reliability in busy washrooms that demand consistency, quality and value.

  • Offers high capacity with each roll
  • Made from 100% recycled fibres
  • European Ecolabeled
  • Unperforated
  • Has a 60 mm core

A quality jumbo roll toilet tissue which offers an efficient solution for high traffic locations with reliable flushability to prevent blockages.
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR