Drukuj
Genomic DNA purification kit, Fermentas
Dostawca: Thermo Fisher Scientific
K0512
K0512EA 958 PLN
K0512
Genomic DNA purification kit, Fermentas
Odczynniki do kwasów nukleinowych Odczynniki i zestawy do oczyszczania kwasów nukleinowych
DNA purification kit is a simple and rapid system for high quality genomic DNA purification from various sources including: whole blood, serum, cell lines, bacterial cells, plant, and mammalian tissues.
  • Efficient - 2 to 10 µg of genomic DNA from 0,2 ml of blood
  • Fast - procedure takes approximately 20 minutes
  • Universal - purifies genomic DNA from various sources
  • Flexible - easy to scale up and down
  • Safe - does not involve phenol extraction
High molecular weight genomic DNA purified with the kit is suitable for direct use in all common molecular biology applications: PCR, FastDigest and conventional restriction digestion, cloning, DNA sequencing, and southern blot analysis.
The kit is based on selective detergent-mediated DNA precipitation from crude lysate. The entire procedure is rapid only 20 to 25 minutes with a typical yield of 2 to 10 µg genomic DNA from 0,2 ml of blood.
Applications include purification of high-quality genomic DNA from wWhole blood or serum, cell culture, plant tissues, mammalian tissues, epithelium samples, bacteria includes lysis solution precipitation solution NaCl solution detailed protocol.
Dodatkowe informacje: The kit contains lysis solution (40 ml), precipitation solution (10X) (8 ml), Sodium chloride (1,2 M) (10 ml). Store the kit at room temperature.
Uwaga: For research use only. Not for use in diagnostic procedures.