Drukuj
Jenway® pH Electrodes
Dostawca: Cole-Parmer
99960-41 68006-40 59530-70 68006-37 99960-34 10090-75 99960-39 99960-44 90269-17
JENW99960-41EA 1060 PLN
JENW99960-41 JENW68006-40 JENW59530-70 JENW68006-37 JENW99960-34 JENW10090-75 JENW99960-39 JENW99960-44 JENW90269-17
Jenway® pH Electrodes
Elektrody Elektrody pH
A variety of pH electrodes for your Jewnay meters.
  • Choose the right electrode for your application
These electrodes are designed for standard measurements of general solutions. The epoxy body makes it especially suitable for field measurements or use in education.