Drukuj
Iridium AAS Standards, (CRMs), Fluka™
Dostawca: Honeywell Chemicals
E4036-100ML E4036-500ML
HONCE4036-100MLEA 1060 PLN
HONCE4036-100ML HONCE4036-500ML
Iridium AAS Standards, (CRMs), Fluka™
Standardy Wzorce kationowe Iridium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certyfikaty: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable