Drukuj
Bromide standard for ion chromatography, (CRM), Fluka™
Dostawca: Honeywell Chemicals
E3006-100ML
HONCE3006-100MLEA 274 PLN
HONCE3006-100ML
Bromide standard for ion chromatography, (CRM), Fluka™
Standardy Wzorce anionowe Bromide Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Certyfikaty: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).