Drukuj
Campylo Thioglycollate Medium Base Dehydrated Culture Media
Dostawca: HIMEDIA
M908-500G
HIMEM908-500GEA 624 PLN
HIMEM908-500G
Campylo Thioglycollate Medium Base Dehydrated Culture Media
Media do mikrobiologii Podłoża sypkie do mikrobiologii
For the isolation, maintenance, and transport of cultures of Campylobacter species.
  • Microbiological culture media
  • 500 g poly bottles
New Product
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable