Drukuj
Avantor® Hichrom Prevail™ Kwas organiczny, kolumny HPLC
Dostawca: Avantor
HICH88648EA 6680 PLN
HICH88648 HICH88655 HICH50755 HICH88640 HICH88645 HICH88745 96429/N
Avantor® Hichrom Prevail™ Kwas organiczny, kolumny HPLC
Kolumny chromatograficzne
Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases. All phases, apart from Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes.
  • Highly efficient silica-based, acid-stable phase
  • Stable from highly organic to highly aqueous
  • Speciality phases for specific applications
  • Excellent sensitivity with microbore and ELSD
Więcej informacji o produkcie