Drukuj
Komora natryskowa
Dostawca: Agilent
G1820-65337
HEWLG1820-65337EA 0 PLN
HEWLG1820-65337
Komora natryskowa
Atomizery
This spray chamber made of quartz is used with 7500 ICP-MS.
It is dedicated for babington, concentric and micro-concentric nebulisers.
Uwaga: Not suitable for hydrofluoric acid.