Drukuj
Sealable Cells for Fluorescence Measurements
Dostawca: HELLMA
117204F-10-40
HELL117204F-10-40EA 3270 PLN
HELL117204F-10-40
Sealable Cells for Fluorescence Measurements
Kuwety Kuwety spektrofotometryczne
Hellma® high precision quartz glass semi-micro cells for fluorescence measurements in the UV/Vis range, 10×4mm path length, with closed screw cap.
  • Cell made of high performance quartz glass
  • Wavelength range from 200 to 2500 nm
  • With closed screw cap and PTFE seal
Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers.
Highest precision: Hellma® precision cells are characterized by the highest precision in path length accuracy and parallelism. This guarantees continuously reproducible measurement