Drukuj
Brush for butyrometer
Dostawca: GLASWARENFABRIK KARL HECHT
91120000012
HECH91120000012EA 996 PLN
HECH91120000012
Brush for butyrometer
Szczotki laboratoryjne
Brush with natural bristles and two brush parts, outer 22×12 mm, inner 60×45 mm. Galvanized iron thread handle.