Drukuj
Pomoc do pipet, Micro-Pipex
Dostawca: GLASWARENFABRIK KARL HECHT
40558001
HECH40558001EA 286 PLN
HECH40558001
Pomoc do pipet, Micro-Pipex
Pipety Kontrolery pipet
Aid for pipetting liquids by using disposable micropipettes with ring mark as well as other pipettes up to 1 ml, e.g. RBC and WBC pipettes.
  • Contaminated disposable micropipettes up to 50 µl are released contactless by the integrated ejection
  • Autoclavable at +121 °C
  • Pipette holder: max. 5 mm Ø
Please note: The pipetting aid may not be used for capillaries end-to-end.
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable