Drukuj
Pudełka do przechowywania i transportu, DURAporter
Dostawca: HEATHROW
HS120052 HS120077 HS120078
HEATHS120052EA 838 PLN
HEATHS120052 HEATHS120077 HEATHS120078
Pudełka do przechowywania i transportu, DURAporter
Pudełka Pudełka transportowe
PC, transparent.
  • Silicone water-tight seal and three lid clasps provide secure closure
  • Lid opens a full 180°, enabling total access to contents and making it easier to clean
  • Will accommodate 13 and 16 mm tubes in a 72-place rack
  • In compliance with U.S. standards, OSHA 29 CFR 1910.1030 and meets UN3373 compliant transport unit requirements
The DURAporter™ is ideal for transporting biological and clinical samples, instruments and products that needs to be kept clean and dry under testing environmental conditions. The carry handle folds neatly into theWięcej informacji o produkcie