Drukuj
Zbiorniki odczynników
Dostawca: HEATHROW
120644 120645 120648 120649 120690 120652 120651 120646 120647 120639 120640 120638 120642 120650 120637 120641 120643
HEAT120644EA 726 PLN
HEAT120644 HEAT120645 HEAT120648 HEAT120649 HEAT120690 HEAT120652 HEAT120651 HEAT120646 HEAT120647 HEAT120639 HEAT120640 HEAT120638 HEAT120642 HEAT120650 613-6915 HEAT120641 HEAT120643
Zbiorniki odczynników
Zbiorniki na odczynniki
These reagent reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures. They are designed for multi-channel pipettes and accommodate most common brands. A full range of volumes and styles is available to accommodate required volume of reagents and reduce waste.
  • 'V' shaped well design allows maximum reagent recovery
  • Easy to read graduation markings located on the top edge with a fill line on inner walls to indicate volume levels
  • Drip-free pour-off spouts at every corner for spill-free pouring
  • A wide base ensures
Więcej informacji o produkcie