Drukuj
Szalki petriego, kwadratowe, Corning® Gosselin™
Dostawca: Corning
BP124-05 BP124-04
GOSSBP124-05EA 1240 PLN
GOSSBP124-05 391-0444
Szalki petriego, kwadratowe, Corning® Gosselin™
Szalki do hodowli Szalki Petriego
PS. Petri plates manufactured under aseptic cleanroom conditions.
  • Manufactured from optically clear virgin polystyrene
  • Sterilised by irradiation to SAL 10⁻³
  • Supplied with 4 vents to provide consistent gas exchange
  • New design to enhance stacked dish stability
Corning® Gosselin™ square petri dishes are optically clear, with a design that has been optimised for use on automated platforms.
The square design is used in colony picking and antibiogram applications.