Drukuj
Bezpieczna stacja aspiracyjna
Dostawca: GILSON
F110744 F110746
GILSF110744EA 9720 PLN
GILSF110744 GILSF110746
Bezpieczna stacja aspiracyjna
Pompy Systemy zasysania próżniowego
The Gilson® safe aspiration station is a stand-alone motorised vacuum system for quick and easy aspiration of biological liquid waste disposal. Liquids are quickly and safely aspirated and collected in a 4 L polypropylene bottle. The station offers adjustable aspiration speed and handles, ideal for ELISA tests, cell culture, and any other contaminated or hazardous liquid aspiration and disposal.
  • Safe and contamination-free
  • Adaptable
  • Convenient
The closed, stand-alone unit is designed to prevent contamination hazards. All parts in contact with liquidsWięcej informacji o produkcie