Drukuj
Restriction enzymes, PstI, FastDigest™, Fermentas
Dostawca: Thermo Fisher Scientific
FD0614 FD0615
FD0614EA 405 PLN
FD0614 FD0615
Restriction enzymes, PstI, FastDigest™, Fermentas
Odczynniki do kwasów nukleinowych Enzymy restrykcyjne
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
  • 100% activity of all FastDigest™ enzymes in one universal reaction buffer
  • Direct loading on gels with FastDigest™ Green buffer
  • 100% buffer compatibility with downstream applications
Designed to complete digestion in 5 to 15 minutes.
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable
Aplikacja
Nazwa enzymu PstI
Źródło
Overhang type 3' overhang
Kompatybilny bufor FastDigest Green Buffer or FastDigest Buffer (colorless)