Drukuj
Restriction enzymes, NcoI, FastDigest™, fermentas
Dostawca: Thermo Fisher Scientific
FD0573 FD0574 FD0575
FD0573EA 138 PLN
FD0573 FD0574 FD0575
Restriction enzymes, NcoI, FastDigest™, fermentas
Odczynniki do kwasów nukleinowych Enzymy restrykcyjne
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
  • 100% activity of all FastDigest™ enzymes in one universal reaction buffer
  • Designed to complete digestion in 5 to 15 minutes
  • Direct loading on gels with FastDigest™ Green buffer
  • 100% buffer compatibility with downstream applications
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable
Aplikacja
Nazwa enzymu FastDigest NcoI
Źródło
Overhang type 5' overhang
Kompatybilny bufor FastDigest Green Buffer or FastDigest Buffer (colorless)