Safemate ECO+ Cabinets are supplied in four different sizes (0.9, 1.2, 1.5 and 1.8). These last generation Microbiological Safety Cabinets Class II type A2, have been certified according to the most stringent safety standards (EN 12469-2000). The internal design, the air flow aerodynamics and monitoring, the built-in safety devices and the very accurate manufacturing, guarantees the highest performances at the most stringent safety levels, as specified by EN 12469 standard and have been certified by the most prestigious European certification bodies for Safety Cabinets.
Safemate ECO+ Cabinets are supplied in four different sizes (0.9, 1.2, 1.5 and 1.8). These last generation Microbiological Safety Cabinets Class II type A2, have been certified according to the most stringent safety standards (EN 12469-2000). The internal design, the air flow aerodynamics and monitoring, the built-in safety devices and the very accurate manufacturing, guarantees the highest performances at the most stringent safety levels, as specified by EN 12469 standard and have been certified by the most prestigious European certification bodies for Safety Cabinets.
- Low energy consumption DC motor blower
- State of the art microprocessor control system, controlled DC motor blower, with volumetric sensor for exhausted air flow monitoring
- Large digital display, high resolution
- Air and aerosol-tight sliding sash, electrically operated by finger touch
- Alarms for low air flow and wrong front window position
- Sloped front and back wall for the most comfortable access
- Comfortable lateral side windows, electrically operated sliding multilayer safety glass window
- Front access for filter maintenance and service
- C-shaped support stand for the easiest one-man installation procedure
- Easy retrofit option kits
Certified intrinsic biological safety, combined with impressively competitive prices, gives the end user a state-of-the-art cabinet accessible to every budget, that only experienced European design and accurate quality manufacturing. The ECO+ series evolves from our best seller series with a more eco-friendly approach: the new DC Motorblowers provide high efficiency while consuming less energy and the airflows has been designed in order to reduce noise pollution, while assuring the high level of operator, product and environment protection required by the EN 12469-2000 standards.
Microprocessor control system offering: Large screen monitor; Automatic control of preset airflow volumes; Sliding sash window with smart control; Permanent monitoring of HEPA filters life span; Alarms. Multilevel alarms, with redundancy functions; Permanent display of working conditions; Highest air flow stability both in case of transitional disturbances or to progressive filter clogging; Continuous monitoring of front barrier air flow for the highest operator safety; Low barrier alarm; Power failure alarm.
Volt-free contact for remote monitoring of exhaust fan. Automatic reset of initial conditions in case of power failure. C-shaped support stand for the easiest one-man installation procedure. Both exhaust and Main Filters are equipped with a micromesh membrane located downstream which acts as airspeed equalizer expansion plenum, as well as a clear indicator of filter damages.
Main structure: cold rolled steel, epoxy powder coated; Stainless steel internal surfaces with SB finishing (including spillage tray). Solid or perforated work surface (300 mm sectors) and special designed front grill in stainless steel (AISI 304 or AISI 316). Comfortable 200 mm front opening.
H14 class High Efficiency Particulate Air filters with 99,999% efficiency on 0,3 μm particles (most penetrating particle diameter) (Efficiency ≥99,995% on 0,1 to 0,2 μm particles MPPS as per EN 1822-1); ISO 3 (ISO14644-1) internal cleanliness level
Mniej informacji